2-Methyl-1,3-benzoxazole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A209252-1G A209252-100MG A209252-10G A209252-250MG
103817-924EA 22 USD
103817-924 103848-612 77086-952 77275-122 77275-124 77275-126
2-Methyl-1,3-benzoxazole-6-carboxylic acid 95%
2-Methyl-1,3-benzoxazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787266
CAS Number: 13452-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR