2-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A112249-100MG A112249-250MG A112249-1G A112249-5G
103797-988EA 118 USD
103797-988 103823-824 77160-790 77160-792 77310-626 77310-628
2-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 95%
2-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
Boiling Pt: 343 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05667936
CAS Number: 54329-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR