1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazole-4-boronic acid, pinacol ester 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]propan-2-ol, 2-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)propan-2-ol, 2-[1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A228632-1G A228632-100MG A228632-250MG A228632-5G A228632-50MG
103819-406EA 59 USD
103819-406 77089-110 77165-474 77165-476 77683-665 77892-371
1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazole-4-boronic acid, pinacol ester 97%, Ambeed.Inc
1-(2-Hydroxy-2-methylprop-1-yl)-1H-pyrazole-4-boronic acid, pinacol ester

Precaución: For research use only.

Formula: C₁₃H₂₃BN₂O₃
Storage Temperature: Freezer
MDL Number: MFCD22378011
CAS Number: 1082503-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR