2-Methyl-1,8-naphthyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1,8-diazanaphthalene, 2-Methylpyrido[2,3-b]pyridine, 2-Methyl[1,8]-Naphthyridine

A210516-100G A210516-250MG A210516-1G A210516-25G A210516-10G
103818-010EA 17 USD
103818-010 103848-706 77012-936 77012-938 77087-078 77087-080 77550-354
2-Methyl-1,8-naphthyridine 98%
2-Methyl-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
Flash Pt: 106.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04066717
CAS Number: 1569-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR