2-Methyl-[1,1'-biphenyl]-3-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A297581-10G
A297581-250MG
A297581-1G
103814-142EA
63
USD
103814-142
103835-284
76648-058
76648-060
76837-970
2-Methyl-[1,1'-biphenyl]-3-carbaldehyde 95%
2-Methyl-[1,1'-biphenyl]-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₂O MW: 196.24 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11553605 CAS Number: 89951-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...