2-Methoxyquinolin-7-amine 97%

Supplier: Ambeed
Warning

A819644-100MG A819644-250MG A819644-1G
103841-126EA 1236 USD
103841-126 77244-316 77244-318 77308-384
2-Methoxyquinolin-7-amine 97%
2-Methoxyquinolin-7-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
Boiling Pt: 335.3±22.0 °C (760 mm Hg)
Flash Pt: 156.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22394145
CAS Number: 1354222-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR