2-Methoxypyrimidine-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A115298-100MG A115298-250MG A115298-5G A115298-1G A115298-25G
103798-540EA 20 USD
103798-540 103824-394 76869-972 76869-974 77297-360 77311-204 77592-244
2-Methoxypyrimidine-5-carbaldehyde 97%
2-Methoxypyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
Boiling Pt: 281.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03446732
CAS Number: 90905-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR