4-Amino-2-methoxypyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxypyrimidin-4-amine

A496865-10G A496865-250MG A496865-25G A496865-1G A496865-5G
103828-262EA 35 USD
103828-262 103839-340 76774-460 76774-462 76774-464 76843-306 76843-308
4-Amino-2-methoxypyrimidine 95%
4-Amino-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.13 g/mol
Boiling Pt: 294.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00128201
CAS Number: 3289-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR