3-Amino-2-methoxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxypyridin-3-amine, 2-Methoxy-3-pyridylamine

A185268-1G A185268-25G A185268-5G A185268-100G A185268-10G A185268-250MG
103815-838EA 9 USD
103815-838 103846-406 77084-042 77084-044 77084-046 77111-564 77111-566 77111-568
3-Amino-2-methoxypyridine 97%
3-Amino-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 240.5 °C (760 mm Hg)
Melting Pt: 67 °C
Flash Pt: 99.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00833386
CAS Number: 20265-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR