2-Methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxybenzeneboronic acid, (2-Methoxyphenyl)metaboric acid, Dihydroxy(2-methoxyphenyl)borane

A109135-100G A109135-10G A109135-1G A109135-25G A109135-500G A109135-5G
103823-242EA 7 USD
103823-242 77135-874 77135-876 77135-878 77135-880 77135-882 77283-810
2-Methoxyphenylboronic acid 98%
2-Methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Boiling Pt: 319.3 °C (760 mm Hg)
Melting Pt: 106…111 °C at N/A mmHg
Flash Pt: 146.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00236047
CAS Number: 5720-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR