(2-Methoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxyphenylacetic acid

A101406-100G A101406-10G A101406-25G A101406-500G A101406-1000G
103807-790EA 147 USD
103807-790 76869-144 76869-146 76869-148 76869-150 77282-718
(2-Methoxyphenyl)acetic acid 97%
(2-Methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄OCH₃)CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 332
Melting Pt: 122…125 °C
Flash Pt: 117.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004321
CAS Number: 93-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR