2-Ethynylanisole 97%

Supplier: Ambeed
Warning

Synonyms: 2'-Methoxyphenyl acetylene, 1-Ethynyl-2-methoxybenzene

A130640-10G A130640-250MG A130640-25G A130640-5G A130640-1G
103801-446EA 11 USD
103801-446 77258-432 77258-434 77298-414 77298-416 77313-898
2-Ethynylanisole 97%
2-Ethynylanisole

Precaución: for Research Use Only

Formula: C₉H₈O
MW: 132.1600 g/mol
Boiling Pt: 204.9 °C (760 mm Hg)
Flash Pt: 91.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03839989
CAS Number: 767-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR