3-Formyl-2-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxynicotinaldehyde, 2-Methoxy-3-pyridinecarboxaldehyde, 2-Methoxy-pyridine-3-carbaldehyde, 2-Methoxypyridine-3-carboxaldehyde

A199771-1G A199771-25G A199771-5G A199771-10G
103817-132EA 12 USD
103817-132 103847-758 77085-806 77085-808 77085-810 77139-304
3-Formyl-2-methoxypyridine 98%
3-Formyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 200…201 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115112
CAS Number: 71255-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR