2-Methoxyisonicotinic acid 98%

Supplier: Ambeed
Warning

A156601-100G A156601-10G A156601-1G A156601-25G A156601-5G
103804-908EA 5 USD
103804-908 103843-108 76768-380 76768-382 77300-202 77318-902 77318-904
2-Methoxyisonicotinic acid 98%
2-Methoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Melting Pt: 198…200 °C
Flash Pt: 185.4 °C
Storage Temperature: Ambient
MDL Number: MFCD04115718
CAS Number: 105596-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR