4-Formyl-2-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxypyridine-4-carbaldehyde, 2-Methoxyisonicotinaldehyde

A160818-250MG A160818-1G A160818-25G A160818-5G A160818-10G
103805-424EA 12 USD
103805-424 103843-658 76812-724 77319-612 77319-614 77319-616 77592-958
4-Formyl-2-methoxypyridine 98%
4-Formyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 234.6 °C (760 mm Hg)
Melting Pt: 32…36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06410680
CAS Number: 72716-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR