2-Methoxyethyl methanesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: Methanesulphonic Acid 2-Methoxyethyl Ester

A101135-25G A101135-5G
103795-816EA 10 USD
103795-816 103821-504 76821-230 77296-992
2-Methoxyethyl methanesulfonate 95%
2-Methoxyethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₄H₁₀O₄S
MW: 154.18 g/mol
Flash Pt: 110.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02656401
CAS Number: 16427-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR