2-Methoxyethyl-p-toluenesulfonate 97%
Supplier: Ambeed
|
Warning
|
A146499-100G
A146499-10G
A146499-500G
A146499-1G
A146499-25G
A146499-5G
103803-674EA
9
USD
103803-674
103841-766
77259-026
77259-028
77259-030
77317-088
77317-090
77317-092
2-Methoxyethyl-p-toluenesulfonate 97%
2-Methoxyethyl-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₄S Boiling Pt: 141 °C (760 mm Hg) Melting Pt: 10 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025910 CAS Number: 17178-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...