2-Methoxyethyl-(2E)-2-acetyl-3-(3-nitrophenyl)acrylate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxyethyl 2-(3-Nitrobenzylidene)-3-oxobutyrate

A448790-5G A448790-100G A448790-25G
103838-846EA 6 USD
103838-846 76842-658 76979-478 76979-480
2-Methoxyethyl-(2E)-2-acetyl-3-(3-nitrophenyl)acrylate 95%
2-Methoxyethyl-(2E)-2-acetyl-3-(3-nitrophenyl)acrylate

Precaución: For research use only

Formula: C₁₄H₁₅NO₆
MW: 293.28 g/mol
Boiling Pt: 433.4 °C (760 mm Hg)
Melting Pt: 72…74 °C
Flash Pt: 182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00521048
CAS Number: 39562-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR