2-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

A117609-10G A117609-25G A117609-1G A117609-5G A117609-100G
103798-912EA 6 USD
103798-912 103824-800 77245-982 77245-984 77311-582 77311-584 77571-056
2-(Methoxycarbonyl)phenylboronic acid 97%
2-(Methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
Boiling Pt: 359 °C (760 mm Hg)
Melting Pt: 90…96 °C
Flash Pt: 170.9 °C
Storage Temperature: Freezer
MDL Number: MFCD02179452
CAS Number: 374538-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR