2-Methoxyethanimidamide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A151005-10G
A151005-250MG
A151005-25G
A151005-1G
A151005-5G
103804-226EA
24
USD
103804-226
103842-372
77259-250
77259-252
77299-848
77317-908
77317-910
2-Methoxyethanimidamide hydrochloride 97%
2-Methoxyethanimidamide hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₉ClN₂O Boiling Pt: 172.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08752294 CAS Number: 1903-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...