2-Methoxy-6-methylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A135671-250MG A135671-1G A135671-5G
103802-202EA 59 USD
103802-202 77027-298 77315-046 77315-048
2-Methoxy-6-methylpyridin-4-amine 95%
2-Methoxy-6-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.167 g/mol
Boiling Pt: 275.2 °C (760 mm Hg)
Flash Pt: 120.2 °C
Storage Temperature: Ambient
MDL Number: MFCD15529703
CAS Number: 89943-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR