6-Methoxy-o-toluidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-1-methoxy-3-methylbenzene, 6-Methyl-o-anisidine, 2-Methoxy-6-methylaniline, 6-Methoxy-o-toluidine, 2-Amino-3-methylanisole, 2-Amino-3-methoxytoluene, 1-Amino-2-methoxy-6-methylbenzene
A102190-100G
A102190-10G
A102190-25G
A102190-1G
A102190-5G
103796-334EA
6
USD
103796-334
103822-028
77257-276
77257-278
77257-280
77308-904
77308-906
6-Methoxy-o-toluidine 98%
6-Methoxy-o-toluidine
Precaución: for Research Use Only
Formula:
C₈H₁₁NO MW: 137.18 g/mol Boiling Pt: 268 °C (760 mm Hg) Melting Pt: 26…29 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075455 CAS Number: 50868-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...