6-Methoxy-2-pyridineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methoxypyridine-2-boronic acid pinacol ester

A407062-1G A407062-5G A407062-250MG
103827-148EA 30 USD
103827-148 103838-188 76841-678 76841-680 77033-712
6-Methoxy-2-pyridineboronic acid pinacol ester 95%
6-Methoxy-2-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₃
Boiling Pt: 329.1±27.0 °C (760 mm Hg)
Melting Pt: 68…72 °C
Flash Pt: 152.8 °C
Storage Temperature: Freezer
MDL Number: MFCD06798266
CAS Number: 1034297-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR