2-Methoxy-5-pyridinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxynicotinic acid

A244590-100G A244590-10G A244590-500G A244590-5G
103851-424EA 6 USD
103851-424 76771-494 76771-496 76771-498 77090-634
2-Methoxy-5-pyridinecarboxylic acid 98%
2-Methoxy-5-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.1400 g/mol
Boiling Pt: 283.8 °C (760 mm Hg)
Flash Pt: 125.4 °C
Storage Temperature: Ambient
MDL Number: MFCD04115721
CAS Number: 66572-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR