2-Methoxy-5-methylpyridin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A102069-10G
A102069-250MG
A102069-1G
A102069-5G
103796-280EA
58
USD
103796-280
103821-976
77135-534
77135-536
77308-860
77308-862
2-Methoxy-5-methylpyridin-4-amine 97%
2-Methoxy-5-methylpyridin-4-amine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD18250750 CAS Number: 1260663-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...