5-(Trifluoromethyl)-o-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-(trifluoromethyl)aniline, 5-(Trifluoromethyl)-2-anisidine, 5-(Trifluoromethyl)-2-methoxyaniline, 2-Amino-4-(trifluoromethyl)anisole, 3-Amino-4-methoxybenzotrifluoride

A188750-5G A188750-100G A188750-10G A188750-500G A188750-1G
103816-132EA 5 USD
103816-132 103846-724 77084-476 77248-496 77248-498 77248-500 77302-152
5-(Trifluoromethyl)-o-anisidine 98%
5-(Trifluoromethyl)-o-anisidine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 230.1 °C (760 mm Hg)
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00042486
CAS Number: 349-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR