2-Methoxy-5-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methoxypyridine-3-boronic acid, pinacol ester, 2-Methoxypyridine-5-boronic acid, pinacol ester

A120696-10G A120696-250MG A120696-25G A120696-5G A120696-1G
103799-982EA 19 USD
103799-982 76971-498 76971-500 76971-502 77297-780 77312-092
2-Methoxy-5-pyridineboronic acid pinacol ester 98%
2-Methoxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₃
MW: 235.09 g/mol
Boiling Pt: 321.8 °C (760 mm Hg)
Melting Pt: 47…51 °C
Flash Pt: 102.2 °C
Storage Temperature: Ambient
MDL Number: MFCD05663858
CAS Number: 445264-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR