3-Amino-2-methoxypyridine-5-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester

A160723-10G A160723-250MG A160723-1G A160723-25G A160723-5G
103805-412EA 10 USD
103805-412 103843-646 76973-576 76973-578 77300-490 77300-492 77300-494
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester 97%
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉BN₂O₃
MW: 250.10 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12923427
CAS Number: 893440-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR