2-Methoxy-4-methylpyridine 98%

Supplier: Ambeed
Danger

A281682-1G A281682-5G A281682-10G
103813-566EA 5 USD
103813-566 103853-516 76837-210 76837-212 77262-666
2-Methoxy-4-methylpyridine 98%
2-Methoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
Boiling Pt: 179.1 °C (760 mm Hg)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD06200799
CAS Number: 100848-70-2
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR