2-Methoxy-4-methylbenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A104374-250MG
A104374-25G
A104374-1G
A104374-5G
103796-698EA
18
USD
103796-698
103822-412
77025-356
77025-358
77309-278
77309-280
2-Methoxy-4-methylbenzaldehyde 97%
2-Methoxy-4-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₁₀O₂ MW: 150.1700 g/mol Boiling Pt: 258 °C (760 mm Hg) Melting Pt: 40…43 °C Flash Pt: 114 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06248007 CAS Number: 57415-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...