2-Methoxy-4-methyl-5-nitropyridine 96%
Supplier: Ambeed
|
Warning
|
A109180-100G
A109180-10G
A109180-250MG
A109180-25G
A109180-1G
A109180-5G
A109180-500G
103797-462EA
6
USD
103797-462
103823-254
76809-844
76809-846
76809-848
76809-850
77310-074
77310-076
77882-916
2-Methoxy-4-methyl-5-nitropyridine 96%
2-Methoxy-4-methyl-5-nitropyridine
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₃ Boiling Pt: 280.3 °C (760 mm Hg) Melting Pt: 77…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095075 CAS Number: 6635-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...