2-Methoxy-4-(4-methylpiperazin-1-yl)aniline 98%

Supplier: Ambeed
Warning

A148730-10G A148730-1G A148730-25G A148730-5G
103803-960EA 8 USD
103803-960 103842-078 77028-012 77317-542 77317-544 77317-546
2-Methoxy-4-(4-methylpiperazin-1-yl)aniline 98%
2-Methoxy-4-(4-methylpiperazin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O
MW: 221.2988 g/mol
Boiling Pt: 392 °C (760 mm Hg)
Flash Pt: 191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11848954
CAS Number: 122833-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR