2-Methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)aniline 95%

Supplier: Ambeed
Warning

A158305-250MG A158305-1G A158305-10G
103805-114EA 12 USD
103805-114 103843-324 76783-716 77319-178 77667-228
2-Methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)aniline 95%
2-Methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₇H₂₈N₄O
MW: 304.43 g/mol
Boiling Pt: 479.3 °C (760 mm Hg)
Melting Pt: 93…97 °C
Flash Pt: 243.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12827955
CAS Number: 761440-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR