2-Methoxy-4-((1-methylpiperidin-4-yl)oxy)aniline 95%

Supplier: Ambeed
Warning

A110504-250MG A110504-1G
103797-674EA 249 USD
103797-674 76809-908 77284-010
2-Methoxy-4-((1-methylpiperidin-4-yl)oxy)aniline 95%
2-Methoxy-4-((1-methylpiperidin-4-yl)oxy)aniline

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MW: 236.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19175279
CAS Number: 761440-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR