2-Methoxy-3-methylpyrazine 98%

Supplier: Ambeed
Danger

A721010-25G A721010-100G
103840-746EA 5 USD
103840-746 76844-962 77006-130
2-Methoxy-3-methylpyrazine 98%
2-Methoxy-3-methylpyrazine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
Boiling Pt: 159.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00006127
CAS Number: 2847-30-5
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR