2-Methoxy-3-methylisonicotinic acid 95%

Supplier: Ambeed
Warning

A320762-100MG A320762-250MG
103825-246EA 373 USD
103825-246 76989-126 76989-128
2-Methoxy-3-methylisonicotinic acid 95%
2-Methoxy-3-methylisonicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 376.9 °C (760 mm Hg)
Flash Pt: 181.7 °C
Storage Temperature: Ambient
MDL Number: MFCD16611502
CAS Number: 1211581-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR