2-Methoxy-3,5-dimethylpyrazine 95%

Supplier: Ambeed
Warning

A109919-250MG A109919-100MG
103797-588EA 67 USD
103797-588 76809-872 77570-554
2-Methoxy-3,5-dimethylpyrazine 95%
2-Methoxy-3,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD00126946
CAS Number: 92508-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR