2-Methoxy-1-naphthaleneboronic acid 98%

Supplier: Ambeed
Warning

A146196-250MG A146196-25G A146196-1G A146196-5G
103803-638EA 6 USD
103803-638 103841-724 76811-996 76811-998 77317-038 77317-040
2-Methoxy-1-naphthaleneboronic acid 98%
2-Methoxy-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
MW: 202.0100 g/mol
Boiling Pt: 425.2±47.0 °C (760 mm Hg)
Flash Pt: 210.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044173
CAS Number: 104116-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR