2'-Methoxybiphenyl-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2'-Methoxy-[1,1'-biphenyl]-4-carbaldehyde, 2'-Methoxy-[1,1'-biphenyl]-4-carboxaldehyde

A101247-250MG A101247-1G
103795-870EA 56 USD
103795-870 103821-554 76780-480 77297-026
2'-Methoxybiphenyl-4-carbaldehyde 97%
2'-Methoxybiphenyl-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 328.7 °C (760 mm Hg)
Flash Pt: 144.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631913
CAS Number: 421553-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR