2-Hydroxyethylmesitylene 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxyethyl)-2,4,6-trimethylbenzene, 2-(2,4,6-Trimethylphenyl)ethanol, 2-Hydroxyethylmesitylene, 2,4,6-Trimethylphenethyl alcohol, 2-Mesitylethanol

A198241-1G A198241-5G
103817-002EA 14 USD
103817-002 103847-628 77302-576 77302-578
2-Hydroxyethylmesitylene 95%
2-Hydroxyethylmesitylene

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 290.7 °C (760 mm Hg)
Flash Pt: 138.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00020609
CAS Number: 6950-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR