2-Mercaptopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Thiopyrimidine

A325029-25G A325029-5G A325029-100G A325029-10G A325029-500G
103836-330EA 9 USD
103836-330 76839-252 76839-254 77263-498 77263-500 77263-502
2-Mercaptopyrimidine 98%
2-Mercaptopyrimidine

Precaución: for Research Use Only

Formula: C₄H₄N₂S
MW: 112.16 g/mol
Boiling Pt: 191.3±23.0 °C (760 mm Hg)
Melting Pt: 133…135 °C
Flash Pt: 69.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006073
CAS Number: 1450-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR