Pyridine-2(1H)-thione 98%

Supplier: Ambeed
Warning

A181001-25G A181001-5G A181001-100G A181001-500G
103845-944EA 5 USD
103845-944 77083-458 77083-460 77096-836 77096-838
Pyridine-2(1H)-thione 98%
Pyridine-2(1H)-thione

Precaución: for Research Use Only

Formula: C₅H₅NS
MW: 111.16 g/mol
Boiling Pt: 184.9 °C (760 mm Hg)
Melting Pt: 127…130 °C at N/A mmHg
Flash Pt: 65.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00457440
CAS Number: 2637-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR