6-Phenyl-2-thiouracil 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Mercapto-6-phenylpyrimidin-4-ol
A304482-10G
A304482-250MG
A304482-1G
A304482-5G
A304482-25G
103814-418EA
29
USD
103814-418
103835-584
76802-594
76802-596
76838-278
76838-280
77305-902
6-Phenyl-2-thiouracil 95%
6-Phenyl-2-thiouracil
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂OS Boiling Pt: 462.3 °C (760 mm Hg) Flash Pt: 233.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059166 CAS Number: 36822-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...