Isopropylmalonic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Isopropylmalonic acid
A465826-100G
A465826-10G
A465826-1G
A465826-25G
A465826-5G
103827-974EA
17
USD
103827-974
103839-048
77291-822
77291-824
77307-440
77307-442
77307-444
Isopropylmalonic acid 95%
Isopropylmalonic acid
Precaución: for Research Use Only
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 315.4 °C (760 mm Hg) Melting Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021717 CAS Number: 601-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...