2-Isopropylaniline 95%

Supplier: Ambeed
Warning

A318434-25G A318434-5G A318434-100G A318434-500G
103836-104EA 9 USD
103836-104 76838-960 76838-962 77263-364 77263-366
2-Isopropylaniline 95%
2-Isopropylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 112…113 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007720
CAS Number: 643-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR