2-Iodothiophene-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A433323-100MG A433323-250MG
103827-550EA 399 USD
103827-550 103838-616 77016-608 77016-610
2-Iodothiophene-3-carbaldehyde 95%
2-Iodothiophene-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₃IOS
MW: 238.05 g/mol
Boiling Pt: 278.9±25.0 °C (760 mm Hg)
Melting Pt: 68…70 °C
Flash Pt: 122.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426915
CAS Number: 18812-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR