2-Iodothiophene 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A277687-1000G A277687-25G A277687-100G A277687-10G A277687-500G A277687-5G
103809-922EA 487 USD
103809-922 103853-296 77093-196 77093-198 77262-580 77262-582 77262-584 77305-264
2-Iodothiophene 98% stabilized
2-Iodothiophene

Precaución: for Research Use Only

Formula: C₄H₃IS
Melting Pt: 40 °C
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00005424
CAS Number: 3437-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR