2-Iodopyrazine 97%

Supplier: Ambeed
Warning

A102067-100G A102067-10G A102067-25G A102067-1G A102067-5G
103796-276EA 5 USD
103796-276 103821-972 77257-268 77257-270 77257-272 77282-898 77308-854
2-Iodopyrazine 97%
2-Iodopyrazine

Precaución: for Research Use Only

Formula: C₄H₃IN₂
MW: 205.99 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319019
CAS Number: 32111-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR