2-Iodonaphthalene 98%
Supplier: Ambeed
|
Warning
|
A307887-100MG
A307887-10G
A307887-250MG
A307887-1G
A307887-25G
A307887-5G
A307887-100G
103814-530EA
11
USD
103814-530
103835-706
76802-702
76802-704
76802-706
76838-452
76838-454
76838-456
77879-618
2-Iodonaphthalene 98%
2-Iodonaphthalene
Precaución: for Research Use Only
Formula:
C₁₀H₇I Boiling Pt: 307.5 °C (760 mm Hg) Melting Pt: 52…56 °C Flash Pt: 138.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021590 CAS Number: 612-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...