2-Iodobenzyl alcohol 97%

Supplier: Ambeed
Warning

A121050-100G A121050-10G A121050-25G A121050-500G A121050-5G
77094-452EA 87 USD
77094-452 77094-454 77094-456 77094-458 77312-140
2-Iodobenzyl alcohol 97%
2-Iodobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 288.3 °C (760 mm Hg)
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00004610
CAS Number: 5159-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR